MC-Val-Cit-PAB-OH ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M3224-100MG
M3224-500MG
TCIAM3224-100MGEA
1700
DKK
TCIAM3224-100MG
TCIAM3224-500MG
MC-Val-Cit-PAB-OH ≥98.0% (by HPLC, total nitrogen)
MC-Val-Cit-PAB-OH
Formel:
C₂₈H₄₀N₆O₇ Opbevaringstemperatur: Fryser |
MDL Number:
MFCD26142966 CAS-nummer: 159857-80-4 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...