Methylamine Acetate ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M3173-1G M3173-5G
TCIAM3173-1GEA 230 DKK
TCIAM3173-1G TCIAM3173-5G
Methylamine Acetate ≥98.0% (by total nitrogen basis)
Methylamine Acetate
Formel: CH₅N·C₂H₄O₂
Kogepunkt: 53°C
CAS-nummer: 6998-30-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR