6-Methoxy-4-chromanone ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
6-Methoxychroman-4-one
M3135-1G
M3135-5G
TCIAM3135-1GEA
879
DKK
TCIAM3135-1G
TCIAM3135-5G
6-Methoxy-4-chromanone ≥97.0% (by GC)
6-Methoxy-4-chromanone
Formel:
C₁₀H₁₀O₃ Kogepunkt: 180°C |
MDL Number:
MFCD00190159 CAS-nummer: 5802-17-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...