1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M3118-1G
M3118-5G
TCIAM3118-1GEA
206
DKK
TCIAM3118-1G
TCIAM3118-5G
1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulphonyl)imide
Formel:
C₁₂H₂₂F₆N₂O₄S₂ Smeltepunkt: 8°C |
CAS-nummer:
380497-17-6 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...