Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M3115-5G M3115-25G
TCIAM3115-5GEA 485 DKK
TCIAM3115-5G TCIAM3115-25G
Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)
Methyl-2-fluoro-5-nitrobenzenecarboxylate
Formel: C₈H₆FNO₄
Kogepunkt: 139°C
MDL Number: MFCD06203689
CAS-nummer: 2965-22-2

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR