4-Methoxy-3-(trifluormethyl)benzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M3095-25G
M3095-5G
TCIAM3095-25GEA
1190
DKK
TCIAM3095-25G
TCIAM3095-5G
4-Methoxy-3-(trifluormethyl)benzoesyre ≥98.0% (by GC, titration analysis)
4-Methoxy-3-(trifluormethyl)benzoesyre
Formel:
C₉H₇F₃O₃ Molekylvægt: 220.15 g/mol |
MDL Number:
MFCD01091014 CAS-nummer: 213598-09-5 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...