2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate ≥95.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acrylic acid 2-[2-(2-Methoxyethoxy)ethoxy]ethyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M3094-100G M3094-500G
TCIAM3094-100GEA 1090 DKK
TCIAM3094-100G TCIAM3094-500G
2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate ≥95.0% (by GC) stabilised
2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate
Formel: C₁₀H₁₈O₅
Molekylvægt: 218.25 g/mol
MDL Number: MFCD30470140
CAS-nummer: 48067-72-7
EINECS: 256-348-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3a33aab4834bc1a7c85d22129f296d4b6339121&vid=f3a33aab4834bc1a7c85d22129f296d4b6339121&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR