Metamizole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonic acid sodium salt, Sulpyrine, Sodium (1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonate, Metamizole sodium salt, Metamizole (sodium salt), Dipyrone
M3060-10G
M3060-50G
TCIAM3060-10GEA
403
DKK
TCIAM3060-10G
TCIAM3060-50G
Metamizole ≥98.0% (by HPLC, titration analysis)
Metamizole
Formel:
C₁₃H₁₆N₃O₄S·Na Smeltepunkt: 227°C |
MDL Number:
MFCD00020783 CAS-nummer: 68-89-3 EINECS: 200-694-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...