Trimethyl(2-methylprop-2-en-1-yl)silane ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Trimethyl(2-methylallyl)silane, Methallyltrimethylsilane

M3010-5ML M3010-25ML
TCIAM3010-5MLEA 624 DKK
TCIAM3010-5ML TCIAM3010-25ML
Trimethyl(2-methylprop-2-en-1-yl)silane ≥95.0% (by GC)
Trimethyl(2-methylprop-2-en-1-yl)silane
Formel: C₇H₁₆Si
Kogepunkt: 112°C
Flammepunkt: –6°C
MDL Number: MFCD00054404
CAS-nummer: 18292-38-1
UN nummer: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39706e4855ed3ab7bed1d4107e946538f263480d&vid=39706e4855ed3ab7bed1d4107e946538f263480d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR