2-Methylquinoline N-Oxide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M3000-1G M3000-5G
TCIAM3000-1GEA 485 DKK
TCIAM3000-1G TCIAM3000-5G
2-Methylquinoline N-Oxide ≥98.0% (by GC)
2-Methylquinoline N-Oxide
Formel: C₁₀H₉NO
CAS-nummer: 1076-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdc5d17a03565cb1bf431d226f627f37fcfbc71f&vid=fdc5d17a03565cb1bf431d226f627f37fcfbc71f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR