5-Methyl-2-phenylpyridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Phenyl-3-picoline

M2969-200MG M2969-1G
TCIAM2969-200MGEA 255 DKK
TCIAM2969-200MG TCIAM2969-1G
5-Methyl-2-phenylpyridine ≥98.0% (by GC)
5-Methyl-2-phenylpyridin
Formel: C₁₂H₁₁N
MDL Number: MFCD04114180
CAS-nummer: 27012-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90128657a1e8261cb262fdfa4fdc2a6e088863bb&vid=90128657a1e8261cb262fdfa4fdc2a6e088863bb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR