α,α,α-Trifluor-p-xylen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M2945-5G
M2945-25G
TCIAM2945-5GEA
731
DKK
TCIAM2945-5G
TCIAM2945-25G
α,α,α-Trifluor-p-xylen ≥98.0% (by GC)
α,α,α-Trifluor-p-xylen
Formel:
C₈H₇F₃ Kogepunkt: 129°C Flammepunkt: 27°C |
MDL Number:
MFCD00075476 CAS-nummer: 6140-17-6 EINECS: 228-124-2 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...