α,α,α-Trifluor-m-xylen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Methyl-3-(trifluoromethyl)benzene, 3-(Trifluoromethyl)toulene, 3-Methylbenzotrifluoride
M2944-1G
M2944-5G
TCIAM2944-1GEA
148
DKK
TCIAM2944-1G
TCIAM2944-5G
α,α,α-Trifluor-m-xylen ≥98.0% (by GC)
α,α,α-Trifluor-m-xylen
Formel:
C₈H₇F₃ Molekylvægt: 160.14 g/mol Kogepunkt: 127 Massefylde: 1.148 Flammepunkt: 27°C |
MDL Number:
MFCD00190105 CAS-nummer: 401-79-6 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...