Methyl (3,5-Dimethoxyphenyl)acetate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M2933-1G M2933-5G
TCIAM2933-1GEA 386 DKK
TCIAM2933-1G TCIAM2933-5G
Methyl (3,5-Dimethoxyphenyl)acetate ≥98.0% (by GC)
Methyl (3,5-Dimethoxyphenyl)acetate
Formel: C₁₁H₁₄O₄
Kogepunkt: 94°C
MDL Number: MFCD00089971
CAS-nummer: 6512-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2add575640e3a6521b08511e0aa9ef0b9311b1da&vid=2add575640e3a6521b08511e0aa9ef0b9311b1da&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR