(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Mesityl(p-tolyl)iodonium trifluoromethanesulphonate, Mesityl(p-tolyl)iodonium triflate
M2909-1G
M2909-5G
TCIAM2909-1GEA
657
DKK
TCIAM2909-1G
TCIAM2909-5G
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate
Formel:
C₁₇H₁₈F₃IO₃S Molekylvægt: 486.29 g/mol |
MDL Number:
MFCD20264882 CAS-nummer: 1204518-02-4 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 177.0 to 182.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...