(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Mesityl(m-tolyl)iodonium trifluoromethanesulphonate, Mesityl(m-tolyl)iodonium triflate

M2908-1G M2908-5G
TCIAM2908-1GEA 821 DKK
TCIAM2908-1G TCIAM2908-5G
(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate
Formel: C₁₇H₁₈F₃IO₃S
MDL Number: MFCD20264883
CAS-nummer: 197245-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a3fad440413d7f3ec26c4be487b868f7158bc1b&vid=0a3fad440413d7f3ec26c4be487b868f7158bc1b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR