Methyl-2-amino-5-fluorobenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M2899-25G M2899-5G
TCIAM2899-25GEA 4230 DKK
TCIAM2899-25G TCIAM2899-5G
Methyl-2-amino-5-fluorobenzoate ≥98.0% (by GC)
Methyl-2-amino-5-fluorobenzoate
Formel: C₈H₈FNO₂
Kogepunkt: 102°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03791308
CAS-nummer: 319-24-4

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR