2-Nitro-5-picoline ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Methyl-2-nitropyridine, 6-Nitro-3-picoline
M2890-1G
M2890-5G
TCIAM2890-1GEA
197
DKK
TCIAM2890-1G
TCIAM2890-5G
2-Nitro-5-picoline ≥98.0% (by GC)
2-Nitro-5-picoline
Formel:
C₆H₆N₂O₂ |
MDL Number:
MFCD00234182 CAS-nummer: 1074-38-0 |
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...