Methyl-6-aminopicolinate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester

M2865-1G M2865-5G
TCIAM2865-1GEA 394 DKK
TCIAM2865-1G TCIAM2865-5G
Methyl-6-aminopicolinate ≥98.0% (by GC)
Methyl-6-aminopicolinate
Formel: C₇H₈N₂O₂
Molekylvægt: 152.1500 g/mol
MDL Number: MFCD00233712
CAS-nummer: 36052-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40bce47e63fe80e9a16919fbb52e83003f6a050b&vid=40bce47e63fe80e9a16919fbb52e83003f6a050b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR