Toluen-α-sulfonylfluorid

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

32395.02 32395.03 32395.04
SERA32395.02EA 246.32 DKK
SERA32395.02 SERA32395.03 SERA32395.04
Toluen-α-sulfonylfluorid
Toluen-α-sulfonylfluorid
Formel: C₇H₇FO₂S
Kogepunkt: 80…85 °C (0,3 mmHg)
Smeltepunkt: 91…92 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007424
CAS-nummer: 329-98-6
EINECS: 206-350-2
UN nummer: 2923
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR