Methyl 5-bromo-2-methyl-3-nitrobenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 5-bromo-2-methyl-3-nitrobenzoate

M2845-5G M2845-1G
TCIAM2845-5GEA 600 DKK
TCIAM2845-5G TCIAM2845-1G
Methyl 5-bromo-2-methyl-3-nitrobenzoate ≥98.0% (by GC)
Methyl-5-bromo-2-methyl-3-nitrobenzoate
Formel: C₉H₈BrNO₄
MDL Number: MFCD07781280
CAS-nummer: 220514-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72022eaa089e433f97d9645a70972af355134c1a&vid=72022eaa089e433f97d9645a70972af355134c1a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR