Methyl 4-Methylcinnamate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M2825-1G M2825-5G
TCIAM2825-1GEA 312 DKK
TCIAM2825-1G TCIAM2825-5G
Methyl 4-Methylcinnamate ≥98.0% (by GC)
Methyl 4-Methylcinnamate
Formel: C₁₁H₁₂O₂
Kogepunkt: 165°C
MDL Number: MFCD00460746
CAS-nummer: 7560-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01a3b7c508fb661ee747a12beab5ecceb550c5af&vid=01a3b7c508fb661ee747a12beab5ecceb550c5af&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR