Mequitazine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 10-(Quinuclidin-3-ylmethyl)-10H-phenothiazine, 10-(1-Azabicyclo[2.2.2]oct-3-ylmethyl)-10H-phenothiazine

M2807-100MG M2807-1G
TCIAM2807-100MGEA 148 DKK
TCIAM2807-100MG TCIAM2807-1G
Mequitazine ≥98.0% (by GC, titration analysis)
Mequitazine
Formel: C₂₀H₂₂N₂S
Molekylvægt: 322.47 g/mol
MDL Number: MFCD01706743
CAS-nummer: 29216-28-2
EINECS: 249-521-7
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,05864
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36c4bfdee5e779c70c767e80ecbd82753f5f2c22&vid=d384c00eaf532b390e4ae1714e8bb1717331d7ac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR