Boc-L-pyroglutamic acid methyl ester ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

M2803-5G M2803-25G
TCIAM2803-5GEA 362 DKK
TCIAM2803-5G TCIAM2803-25G
Boc-L-pyroglutamic acid methyl ester ≥98.0% (by GC)
Boc-L-pyroglutamic acid methyl ester
Formel: C₁₁H₁₇NO₅
Molekylvægt: 243.26 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06809720
CAS-nummer: 108963-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb106cffe904d0c89cdb4e031c9e62fd9484ee6d&vid=bb106cffe904d0c89cdb4e031c9e62fd9484ee6d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Specific rotation [a]20/D -30.0 to -34.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR