2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]benzimidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, Lansoprazole sulphide
M2800-1G
M2800-5G
TCIAM2800-1GEA
312
DKK
TCIAM2800-1G
TCIAM2800-5G
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole
Formel:
C₁₆H₁₄F₃N₃OS Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00834357 CAS-nummer: 103577-40-8 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...