Methyl-2,6-difluoro-3-nitrobenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2,6-difluoro-3-nitrobenzoate

M2749-1G M2749-5G
TCIAM2749-1GEA 132 DKK
TCIAM2749-1G TCIAM2749-5G
Methyl-2,6-difluoro-3-nitrobenzoate ≥98.0% (by GC)
Methyl-2,6-difluoro-3-nitrobenzoate
Formel: C₈H₅F₂NO₄
Kogepunkt: 151°C
MDL Number: MFCD07368850
CAS-nummer: 84832-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feb4575e9b9f44cfca1324417d3a291fa7a78a61&vid=feb4575e9b9f44cfca1324417d3a291fa7a78a61&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR