4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol, 4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine

M2739-1G M2739-5G
TCIAM2739-1GEA 255 DKK
TCIAM2739-1G TCIAM2739-5G
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Formel: C₅H₃F₃N₂OS
Molekylvægt: 196.10 g/mol
MDL Number: MFCD00112978
CAS-nummer: 368-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb157445810845b28ec63d35b0d0a28ac3fd9eab&vid=d5fc782ed7c8cf00fc6626aebc412c985720d909&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 245.0 to 249.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR