Methyl4-ethoxybenzoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M2738-25G M2738-5G
TCIAM2738-25GEA 961 DKK
TCIAM2738-25G TCIAM2738-5G
Methyl4-ethoxybenzoat ≥98.0% (by GC)
Methyl4-ethoxybenzoat
Formel: C₁₀H₁₂O₃
Kogepunkt: 260°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00017264
CAS-nummer: 23676-08-6
EINECS: 245-817-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR