(4-Tolyloxy)eddikesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(p-Tolyloxy)acetic acid, 4-Methylphenoxyacetic acid, (4-Methylphenoxy)acetic acid
M2729-25G
M2729-5G
TCIAM2729-25GEA
2100
DKK
TCIAM2729-25G
TCIAM2729-5G
(4-Tolyloxy)eddikesyre ≥98.0% (by GC, titration analysis)
(4-Tolyloxy)eddikesyre
Formel:
(C₆H₄(CH₃))OCH₂COOH |
MDL Number:
MFCD00014365 CAS-nummer: 940-64-7 EINECS: 213-374-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...