Methyl6-Bromnicotinat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 6-bromonicotinate
M2682-1G
M2682-5G
TCIAM2682-1GEA
140
DKK
TCIAM2682-1G
TCIAM2682-5G
Methyl6-Bromnicotinat ≥98.0% (by GC)
Methyl6-Bromnicotinat
Formel:
C₇H₆BrNO₂ |
MDL Number:
MFCD04972371 CAS-nummer: 26218-78-0 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...