(+)-Menthyl acetate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (+)-Acetoxy-p-menthane

M2631-1ML M2631-5ML
TCIAM2631-1MLEA 452 DKK
TCIAM2631-1ML TCIAM2631-5ML
(+)-Menthyl acetate ≥98.0% (by GC)
(+)-Menthyl acetate
Formel: C₁₂H₂₂O₂
Molekylvægt: 198.31 g/mol
Flammepunkt: 77°C
MDL Number: MFCD00062977
CAS-nummer: 5157-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79d3139aec8344499c9bc636b4309fc3033e42c8&vid=79d3139aec8344499c9bc636b4309fc3033e42c8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 96.0 ee%
Specific rotation [a]20/D +76.0 to +79.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR