2-Morpholinoacetic acid hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Morpholinoacetic acid hydrochloride, Morpholin-4-ylacetic acid hydrochloride, 4-Morpholineacetic acid hydrochloride
M2608-1G
M2608-5G
TCIAM2608-1GEA
337
DKK
TCIAM2608-1G
TCIAM2608-5G
2-Morpholinoacetic acid hydrochloride ≥98.0% (by titrimetric analysis)
2-Morpholinoacetic acid hydrochloride
Formel:
C₆H₁₂ClNO₃ |
MDL Number:
MFCD03540888 CAS-nummer: 89531-58-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...