Methylindol-3-ylacetat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: IAA methyll ester, Methyl indole-3-acetate, Indole-3-acetic acid methyl ester, Indol-3-ylacetic acid methyl ester

M2605-5G M2605-25G
TCIAM2605-5GEA 526 DKK
TCIAM2605-5G TCIAM2605-25G
Methylindol-3-ylacetat ≥98.0% (by GC)
Methylindol-3-ylacetat
Formel: C₁₁H₁₁NO₂
Kogepunkt: 160°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00022749
CAS-nummer: 1912-33-0
EINECS: 217-622-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e2da67e0c1031c1ed9aaf73460c8b97fafd6141&vid=98cda2d233778b1cf535316f05ef7a959e8f4526&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown to Dark green powder to lump
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR