Methyl-2-aminothiazole-5-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester
M2598-1G
M2598-5G
TCIAM2598-1GEA
263
DKK
TCIAM2598-1G
TCIAM2598-5G
Methyl-2-aminothiazole-5-carboxylate ≥98.0% (by GC)
Methyl-2-aminothiazole-5-carboxylate
Formel:
C₅H₆N₂O₂S Molekylvægt: 158.1800 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD03788562 CAS-nummer: 6633-61-0 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 189.0 to 194.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...