5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone, Metaxalone, 5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone

M2578-1G M2578-5G
TCIAM2578-1GEA 288 DKK
TCIAM2578-1G TCIAM2578-5G
5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one ≥98.0% (by GC)
5-((3,5-Dimethylphenoxy)methyl)oxazolidin-2-one
Formel: C₁₂H₁₅NO₃
Molekylvægt: 221.25 g/mol
Kogepunkt: 225°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00867700
CAS-nummer: 1665-48-1
EINECS: 216-777-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1b8588344178c8e86e3e5284e5ad7a1d838a2ad&vid=f1b8588344178c8e86e3e5284e5ad7a1d838a2ad&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR