2-Methylnaphth[2,1-d]oxazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M2559-1G M2559-5G
TCIAM2559-1GEA 296 DKK
TCIAM2559-1G TCIAM2559-5G
2-Methylnaphth[2,1-d]oxazole ≥98.0% (by GC)
2-Methylnaphth[2,1-d]oxazole
Formel: C₁₂H₉NO
Kogepunkt: 302°C
MDL Number: MFCD00022185
CAS-nummer: 20686-65-1
EINECS: 243-971-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1967c19a5d55f44320cc3d8dc127da36c3f9efe&vid=d1967c19a5d55f44320cc3d8dc127da36c3f9efe&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR