Erbstatin analog ≥96.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Erbstatin Analog, 2,5-Dihydroxycinnamic acid methyll ester, Methyl 2,5-Dihydroxycinnamate, Methyl 3-(2,5-dihydroxyphenyl)acrylate

M2520-10MG M2520-100MG
TCIAM2520-10MGEA 403 DKK
TCIAM2520-10MG TCIAM2520-100MG
Erbstatin analog ≥96.0% (by HPLC)
Erbstatin analog
Formel: C₁₀H₁₀O₄
MDL Number: MFCD00132932
CAS-nummer: 63177-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9352de14132ea6cf94a4132e4ec95df25cb8fca&vid=d9352de14132ea6cf94a4132e4ec95df25cb8fca&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 96.0 area%
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR