Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester
M2496-1G
M2496-5G
TCIAM2496-1GEA
517
DKK
TCIAM2496-1G
TCIAM2496-5G
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate ≥98.0% (by GC)
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate
Formel:
C₅H₄F₆O₂ Molekylvægt: 210.08 g/mol Kogepunkt: 88°C |
MDL Number:
MFCD00054663 CAS-nummer: 360-54-3 UN nummer: 1992 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...