2-(4-Methyl-5-thiazolyl)ethyl butyrate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Methylthiazol-5-yl)ethyl butyrate, 5-(2-Butyryloxyethyl)-4-methylthiazole, Butyric Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester

M2489-5G M2489-25G
TCIAM2489-5GEA 206 DKK
TCIAM2489-5G TCIAM2489-25G
2-(4-Methyl-5-thiazolyl)ethyl butyrate ≥98.0% (by GC)
2-(4-Methyl-5-thiazolyl)ethyl butyrate
Formel: C₁₀H₁₅NO₂S
Molekylvægt: 213.30 g/mol
Kogepunkt: 136°C
MDL Number: MFCD09032913
CAS-nummer: 94159-31-6
EINECS: 303-209-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aee08f132b57788728a557f4dce0ad257c7c9ee9&vid=aee08f132b57788728a557f4dce0ad257c7c9ee9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR