Methyl2-Bromnicotinat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2-bromonicotinate, Methyl 2-bromopyridine-3-carboxylate

M2449-1G M2449-5G
TCIAM2449-1GEA 452 DKK
TCIAM2449-1G TCIAM2449-5G
Methyl2-Bromnicotinat ≥98.0% (by GC)
Methyl2-Bromnicotinat
Formel: C₇H₆BrNO₂
MDL Number: MFCD07367915
CAS-nummer: 52718-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c729eb6fe0e7d3452f43e8979f68ef08c6b17d7f&vid=c729eb6fe0e7d3452f43e8979f68ef08c6b17d7f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR