5-Methoxy-1H-indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Methoxyindole-3-carboxylic acid

M2438-1G M2438-5G
TCIAM2438-1GEA 485 DKK
TCIAM2438-1G TCIAM2438-5G
5-Methoxy-1H-indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Methoxy-1H-indole-3-carboxylic acid
Formel: C10H9NO3
Molekylvægt: 191.19 g/mol
MDL Number: MFCD03265451
CAS-nummer: 10242-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e11f5cc7c84535ff2ed55a855c6ef2ef3864eb5&vid=1e11f5cc7c84535ff2ed55a855c6ef2ef3864eb5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 6.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR