Methyl3-methyl-2-furoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 3-methylfuran-2-carboxylate

M2377-1G M2377-5G
TCIAM2377-1GEA 165 DKK
TCIAM2377-1G TCIAM2377-5G
Methyl3-methyl-2-furoat ≥98.0% (by GC)
Methyl3-methyl-2-furoat
Formel: C₇H₈O₃
Molekylvægt: 140.14 g/mol
Kogepunkt: 74°C
MDL Number: MFCD00014110
CAS-nummer: 6141-57-7
EINECS: 228-131-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d723871997b53773f008982062d3f204c35fc9b&vid=5d723871997b53773f008982062d3f204c35fc9b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR