3-Methyl-2-furoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Methylfuran-2-carboxylic acid

M2376-1G M2376-5G
TCIAM2376-1GEA 329 DKK
TCIAM2376-1G TCIAM2376-5G
3-Methyl-2-furoic acid ≥98.0% (by GC, titration analysis)
3-Methyl-2-furoic acid
Formel: C₆H₆O₃
MDL Number: MFCD00014111
CAS-nummer: 4412-96-8
EINECS: 224-571-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e9eecbd9a9f46ed2464cd7ace10e9e48744ec7e&vid=7e9eecbd9a9f46ed2464cd7ace10e9e48744ec7e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR