Mometasone furoate ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M2354-200MG
M2354-1G
TCIAM2354-200MGEA
394
DKK
TCIAM2354-200MG
TCIAM2354-1G
Mometasone furoate ≥98.0% (by HPLC)
Mometasone furoate
Formel:
C₂₇H₃₀Cl₂O₆ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00866003 CAS-nummer: 83919-23-7 Merck Index: 14,06241 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 212.0 to 216.0 deg-C |
Specific rotation [a]20/D | +48.0 to +52.0 deg (C=0.5、MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...