(3-Methoxyphenyl)acetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Methoxybenzyl cyanide, 2-(3-Methoxyphenyl)acetonitrile

M2331-5G M2331-25G
TCIAM2331-5GEA 165 DKK
TCIAM2331-5G TCIAM2331-25G
(3-Methoxyphenyl)acetonitril ≥98.0% (by GC)
(3-Methoxyphenyl)acetonitril
Formel: C₉H₉NO
Molekylvægt: 147.18 g/mol
Kogepunkt: 131°C
Massefylde: 1.05
Flammepunkt: 98°C
MDL Number: MFCD00001910
CAS-nummer: 19924-43-7
EINECS: 243-428-5
UN nummer: 3276
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR