2-Methoxyethyl methacrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethylene Glycol Monomethyl Ether Methacrylate, Methacrylic Acid 2-Methoxyethyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M2282-25G M2282-500G
TCIAM2282-25GEA 132 DKK
TCIAM2282-25G TCIAM2282-500G
2-Methoxyethyl methacrylate ≥98.0% (by GC) stabilised
2-Methoxyethyl methacrylate
Formel: C₇H₁₂O₃
Molekylvægt: 144.17 g/mol
Kogepunkt: 65°C
Flammepunkt: 66°C
MDL Number: MFCD00048122
CAS-nummer: 6976-93-8
EINECS: 230-241-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR