1-Methyl-3-[6-(methylsulphinyl)hexyl]imidazolium p-toluenesulphonate ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M2274-1G
M2274-5G
TCIAM2274-1GEA
1920
DKK
TCIAM2274-1G
TCIAM2274-5G
1-Methyl-3-[6-(methylsulphinyl)hexyl]imidazolium p-toluenesulphonate ≥97.0% (by HPLC, total nitrogen)
Formel:
C₁₈H₂₈N₂O₄S₂ |
CAS-nummer:
1352947-66-0 |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...