Mebendazole ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate, Methyl (5-benzoyl-1H-benzo[d]imidazol-2-yl)carbamate

M2273-25G M2273-5G
TCIAM2273-25GEA 657 DKK
TCIAM2273-25G TCIAM2273-5G
Mebendazole ≥98.0% (by HPLC, total nitrogen)
Mebendazole
Formel: C₁₆H₁₃N₃O₃
MDL Number: MFCD00057872
CAS-nummer: 31431-39-7
EINECS: 250-635-4
Merck Index: 14,05768

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR