Methyl5-Bromvalerat ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 5-Bromovalerate, 5-Bromovaleric acid methyl ester
M2265-5G
M2265-25G
TCIAM2265-5GEA
312
DKK
TCIAM2265-5G
TCIAM2265-25G
Methyl5-Bromvalerat ≥97.0% (by GC)
Methyl5-Bromvalerat
Formel:
BrH₂C(CH₂)₃COOCH₃ Kogepunkt: 89°C Flammepunkt: 99°C |
MDL Number:
MFCD00000265 CAS-nummer: 5454-83-1 EINECS: 226-709-7 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...