Methyl-4-bromo-3-hydroxybenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 4-bromo-3-hydroxybenzoate

M2246-1G M2246-5G
TCIAM2246-1GEA 444 DKK
TCIAM2246-1G TCIAM2246-5G
Methyl-4-bromo-3-hydroxybenzoate ≥98.0% (by GC)
Methyl-4-bromo-3-hydroxybenzoate
Formel: C₈H₇BrO₃
Molekylvægt: 231.0400 g/mol
MDL Number: MFCD08056372
CAS-nummer: 106291-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=144f4e5246e9680a2094007b0d388c3a707af42c&vid=144f4e5246e9680a2094007b0d388c3a707af42c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR